Order Entry
Denmark
ContactUsLinkComponent
Deuterium oxide 99.8 atom % D, AcroSeal® for NMR spectroscopy
Deuterium oxide 99.8 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 426931000.
CAS Number:  
undefined
Deuterium oxide 99.8 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 426931000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • D-Enrichment:
    ≥99.75 %
  • Water:
    ≥30.0 %
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

CAS Number:7789-20-0
 :
Formula:D₂O
MDL Number:MFCD00044636
 :20 g/mol
Boiling Pt:101.4 °C (760 mmHg)
Melting Pt:3.8 °C
Density:1.105 g/cm³
Storage Temperature:Ambient