Order Entry
Denmark
ContactUsLinkComponent
Potassium chloride for biochemistry
Potassium chloride for biochemistry
Certificates
undefined
Potassium chloride for biochemistry
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Argentometric:
    ≥99.0 %
  • Solubility:
    (1 M in water) Clear colorless
  • UV:
    (1 M in water)
  • pH:
    5.5 to 8.0 (5 % in water)
  • Iron (Fe):
    ≤0.0005 %
  • Magnesium (Mg):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together