
2-Imidazolidinone, anhydrous 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120-93-4
:
Formula:C₃H₆N₂O
MDL Number:MFCD00005257
:86.0925 g/mol
Boiling Pt:187 °C (10Torr)
Melting Pt:131.4…132.4 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥95.0 %
- GC:≥99.75 %
- UV:at 280 nm A: ≤0.03
- Color scale:≤10 APHA
- Specific gravity:(20°C) 0.900 to 0.904
- Residue after evaporation:≤0.0002 %
- Acidity:≤0.0008 meq/g
Specification Test Results
Frequently Bought Together