β-Cyclodextrine 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
- CAS Number:7585-39-9
- Formula:C₄₂H₇₀O₃₅
- MDL Number:MFCD00078139
- Molecular Weight:1134.98 g/mol
- Melting Pt:310 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
- CAS Number:7585-39-9
- Formula:C₄₂H₇₀O₃₅
- MDL Number:MFCD00078139
- Molecular Weight:1134.98 g/mol
- Melting Pt:310 °C
- Storage Temperature:Ambient
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤14 % (105°C, 8 hrs)
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:+159° to +165° (20°C, 589 nm) (c=1.5, H2O) (on dry substance)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



