Anthrone 95%, pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Boiling Pt:721 °C (760 mmHg)
- Melting Pt:152…157 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Boiling Pt:721 °C (760 mmHg)
- Melting Pt:152…157 °C
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to beige Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:155 to 161 °C
- GC:≥94 %



