Order Entry
Denmark
ContactUsLinkComponent
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 97%
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 97%
Catalog # ACRO393330010
CAS Number:  
undefined
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 97%
Catalog # ACRO393330010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥96.0 %
  • Specific optical rotation:
    +5° to +7° (20°C, 589 nm) c=5, chloroform

Specification Test Results

About this item

 :
 :262.302 g/mol
Melting Pt:105…114 °C
Storage Temperature:Refrigerator
CAS Number:1707-77-3
Formula:C₁₂H₂₂O₆
MDL Number:MFCD00003211