



Trimethylsulphonium hydroxide 0.25 M in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17287-03-5
:
Formula:C₃H₁₀OS
MDL Number:MFCD00216756
:94.17 g/mol
Density:0.81 g/cm³
Flash Pt:11 °C
UN:3286
ADR:3,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HCl:0.20 to 0.30 M
Specification Test Results
Frequently Bought Together