Order Entry
Denmark
ContactUsLinkComponent
Lithium acetate, anhydrous 99%, extra pure
Irritant
Lithium acetate, anhydrous 99%, extra pure
Certificates
undefined
Irritant
Lithium acetate, anhydrous 99%, extra pure

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    280 to 285 °C
  • Water:
    ≤0.5 %
  • Titration with HClO4:
    ≥98.5 %
  • Insoluble matter:
    ≤0.1 % (in water)

Specification Test Results

Related Information

Frequently Bought Together