Order Entry
Denmark
ContactUsLinkComponent
Lithium nitrate 99.999% (trace metals basis)
Irritant
Oxidizing
Lithium nitrate 99.999% (trace metals basis)
Certificates
undefined
Irritant
Oxidizing
Lithium nitrate 99.999% (trace metals basis)

Specification Test Results

  • Appearance:
    White Crystalline powder and/or chunks
  • Total trace metal impurities:
    ≤10 ppm
  • Lithium (Li):
    9 to 11 %

Specification Test Results

Related Information

Frequently Bought Together