Order Entry
Denmark
ContactUsLinkComponent
2,2-Dimethylpropane-1,3-diamine 99%
2,2-Dimethylpropane-1,3-diamine 99%
Certificates
undefined
2,2-Dimethylpropane-1,3-diamine 99%

Specification Test Results

  • Appearance:
    Colorless Low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4556 to 1.4576 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Titration with HClO4:
    98.5 to 101.5 %

Specification Test Results

Related Information

Frequently Bought Together