Order Entry
Denmark
Orders LinkContactUsLinkComponent
Potassium sulphate, pure

Potassium sulphate, pure

Potassium sulphate, pure
Certificates
Potassium sulphate, pure

3 Options Available Below

Product Details & Documents
  • CAS Number:7778-80-5
  • Formula:K₂SO₄
  • MDL Number:MFCD00011388
  • Molecular Weight:174.26 g/mol
  • Boiling Pt:1689 °C (760 mmHg)
  • Melting Pt:1067 °C
  • Density:2,66 g/cm³ (20 °C)
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1 % (130°C)
  • pH:
    5.5 to 8.5
  • Titration with HClO4:
    ≥99.0 %
  • Arsenic (As):
    ≤0.0003 %
  • Lead (Pb):
    ≤0.0002 %
  • Mercury (Hg):
    ≤0.0001 %
  • Selenium (Se):
    ≤0.0005 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 492,00 kr
    492,00 kr
    1 * 1 kg
       492,00 kr

  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1.240,00 kr
    1.240,00 kr
    1 * 5 kg
       1.240,00 kr

  • Fibre drum
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 3.780,00 kr
    3.780,00 kr
    1 * 25 kg
       3.780,00 kr