Lithium chloride 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Boiling Pt:1382 °C (760 mmHg)
Melting Pt:605 °C
Density:2.06 g/cm³
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals or granules
- Titration Argentometric:≥98.5 %
- Loss on drying:≤0.5 % (400°C, 4 h)
- Insoluble matter:≤0.01 % (in acid)
Specification Test Results
Frequently Bought Together