Order Entry
Denmark
ContactUsLinkComponent
Deuterium oxide 100.0 atom % D, AcroSeal® for NMR spectroscopy
Deuterium oxide 100.0 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 184761000.
CAS Number:  
undefined
Deuterium oxide 100.0 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 184761000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • D-Enrichment:
    ≥99.95 %
  • Melting point:
    143 to 152 °C
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥97.5 %
  • Titration with NaOH:
    18.5 to 19.5 % (Fumaric acid)

Specification Test Results

About this item

CAS Number:7789-20-0
 :
Formula:D₂O
MDL Number:MFCD00044636
 :20 g/mol
Boiling Pt:101.4 °C (760 mmHg)
Melting Pt:3.8 °C
Density:1.105 g/cm³
Storage Temperature:Ambient