Heating thermostats with stainless steel bath, PRO series
Supplier: LAUDA
Certificates
About this item
The PRO heating bath thermostats are optimised for internal bath applications and offer excellent temperature stability. The removable operating units, including mount, allow considerable reduction in the height of the device.
- Touchscreen can be operated with gloves
- Internal Varioflex pump (pressure-suction pump) with eight pump levels
- Programmer with one program, 20 segments or 100 programs, with 50 segments for command unit
- Individual access rights for upto 20 users
- Enables ramp function for easy computation
Units have a 360º turnable and detachable control panel for remote control. High contrast OLED display for units with Base control, whereas large colour touchscreen (145 mm) with multi touch operation for units with Command Touch, with timer and count down display.
Max. flow rate / max. pressure:
Pump pressure/suction: 0,7/0,4 bar
Pump flow pressure/suction: 22/20 L/min
Delivery: Supplied with bath covers and 2 hose olives with cap nuts for the cooling coil.
Frequently Bought Together