Bottles, wide neck, depyrogenated glass
Supplier: EP SCIENTIFIC
Certificates
About this item
Clear or amber glass with closure.
- Packaging and storing articles that will be terminally sterilised
- Storage of laboratory reagents and medias
- Sample storage and water sampling
- Meet endotoxin levels of less than 0,06 EU/ml
- Amber bottles protect light-sensitive materials
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Recommended for endotoxin-free applications
Particle-certified glass containers are tested and certified to meet the low particle criteria; assembled to contain as few as five particles per milliliter >0,5 µm.
Recommended for low particle applications. Individual bottles double bagged to maintain integrity until point of use.
Ordering information: Custom cleaning services for additional applications are available; contact your Avantor sales representative for more information.
Delivery: It is supplied with certificate of analysis.
Specifications
- Catalog No:
- EPSCC20-04A/PF
- EPSC117-2L/PF
- EPSC133-32C/PF
- EPSC117-4L/PF
- EPSC122-16A/PF
- EPSC121-08A/PF
- EPSC123-40A/PF
- EPSC123-32A/PF
- EPSC123-80A/PF
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Frequently Bought Together