Tryptoline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole, Tetrahydro-β-carboline
CAS Number:16502-01-5
:
Formula:C₁₁H₁₂N₂
MDL Number:MFCD00004954
Molecular Weight:172.231 g/mol
Melting Pt:206…208 °C
Storage Temperature:Ambient
UN:3263
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to orange to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together