Order Entry
Denmark
ContactUsLinkComponent
2-Iodoxybenzoic acid stabilised
2-Iodoxybenzoic acid stabilised
Certificates
undefined
2-Iodoxybenzoic acid stabilised
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to beige Powder or crystals
  • Titration Iodometric:
    39 to 50 % as 2-Iodoxybenzoic acid
  • Identification:
    HPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid, Isophthalic acid, Benzoic acid)