Br-Mmc (=4-bromomethyl-7-methoxycoumarin) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35231-44-8
:
Formula:C₁₁H₉BrO₃
MDL Number:MFCD00006869
:269.09 g/mol
Melting Pt:212…217 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to yellow to beige Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:212 to 217 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together