Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
Ruthenium(III) nitrosylnitrate in diluted aq. nitric acid solution
 
undefined
 
 
 
Ruthenium(III) nitrosylnitrate in diluted aq. nitric acid solution

 

  • Appearance:
    Clear orange to dark brown Solution
  • Assay:
    1.5 % Ru (typically)

 

 

Frequently Bought Together