Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
Palladium 5% on calcium carbonate lead poisoned (3.5% Pb)
 
undefined
 
 
 
Palladium 5% on calcium carbonate lead poisoned (3.5% Pb)

 

  • Appearance:
    Grey to grey-black Powder
  • Assay:
    5 % (Pd)
  • Water:
    ≤5.0 %
  • Additional info:
    poisoned with 3.5% lead

 

 

Frequently Bought Together