Order Entry
Denmark
ContactUsLinkComponent
Polydimethylsiloxane, 100 cSt
Polydimethylsiloxane, 100 cSt
Certificates
undefined
Polydimethylsiloxane, 100 cSt

Specification Test Results

  • Appearance:
    Clear viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4020 to 1.4050 (20°C, 589 nm)
  • Color scale:
    ≤35 APHA
  • Viscosity:
    95 to 105 cSt (at 25°C)
  • Volatiles:
    ≤0.5 % (200°C)

Specification Test Results

Related Information

Frequently Bought Together