Cellulose triacetate
Supplier: Thermo Fisher Scientific
About this item
Appearance: White to light yellow Pellets or rods
- CAS Number:9012-09-3
- Formula:C₄₀H₅₄O₂₇
- Molecular Weight:966.845 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Appearance: White to light yellow Pellets or rods
- CAS Number:9012-09-3
- Formula:C₄₀H₅₄O₂₇
- Molecular Weight:966.845 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Pellets or rods
- Infrared spectrum:Conforms
- Acetyl content:43 to 44 wt %
- Titration Argentometric:≥95 % (on dry substance)
- Water:≤10 %



