
7,7,8,8-Tetracyanoquinodimethane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1518-16-7
:
Formula:C₁₂H₄N₄
MDL Number:MFCD00011664
:204.19 g/mol
Melting Pt:287…289 °C (dec.)
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Green to orange-brown Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together