Order Entry
Denmark
ContactUsLinkComponent
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose 99%
Certificates
undefined
2-Deoxy-D-galactose 99%
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to light beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 % (ELSD)
  • Specific optical rotation:
    +56° to +60° (20°C, 589 nm) (c=2, H2O, 5 h)