Order Entry
Denmark
ContactUsLinkComponent
(1S)-1-Phenylpropane-1,3-diol 98%
(1S)-1-Phenylpropane-1,3-diol 98%
Certificates
undefined
(1S)-1-Phenylpropane-1,3-diol 98%

Specification Test Results

  • Appearance:
    White Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    62 to 66 °C
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -53° to -56° (22 °C, 589 nm) (c=1, 1,2-dichlorethane)

Specification Test Results

Related Information

Frequently Bought Together