Order Entry
Denmark
Orders LinkContactUsLinkComponent
Papain
Papain
Certificates
Papain

3 Options Available Below

Product Details & Documents

CAS No.: 9001-73-4

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ACRO416760100
  • ACRO416761000
  • ACRO416765000
  • Enzyme name:
  • Papain
  • Papain
  • Papain
  • MDL No.:
  • MFCD00131791
  • MFCD00131791
  • MFCD00131791
  • CAS No.:
  • 9001-73-4
  • 9001-73-4
  • 9001-73-4
  • Form:
  • Cream to light brown Powder
  • Cream to light brown Powder
  • Cream to light brown Powder
  • Storage conditions:
  • Refrigerator
  • Refrigerator
  • Refrigerator

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 305,00 kr
    305,00 kr
    1 * 10 g
       305,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO416760100
    Enzyme name
    Papain
    MDL No.
    MFCD00131791
    CAS No.
    9001-73-4
    Form
    Cream to light brown Powder
    Storage conditions
    Refrigerator
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.470,00 kr
    1.470,00 kr
    1 * 100 g
       1.470,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO416761000
    Enzyme name
    Papain
    MDL No.
    MFCD00131791
    CAS No.
    9001-73-4
    Form
    Cream to light brown Powder
    Storage conditions
    Refrigerator
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 5.590,00 kr
    5.590,00 kr
    1 * 500 g
       5.590,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO416765000
    Enzyme name
    Papain
    MDL No.
    MFCD00131791
    CAS No.
    9001-73-4
    Form
    Cream to light brown Powder
    Storage conditions
    Refrigerator
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.