Aluminum phosphate, pure
:7784-30-7
:
:AlPO₄
:MFCD00003430
:121.951 g/mol
:>1500 °C
:Ambient
- Appearance:White Powder
- Titration:≥80.0 % (AlPO4)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Special pipette tips, Finntip™
: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals