Hexadecyl palmitate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-10-3
:
Formula:C₃₂H₆₄O₂
MDL Number:MFCD00053739
Boiling Pt:360 °C (760 mmHg)
Melting Pt:52…56 °C
:480.86 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:52 to 56 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together