Order Entry
Denmark
ContactUsLinkComponent
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 99%
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 99%
Certificates
undefined
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 99%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    49 to 53 °C
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +57° to +61° (20 °C, 589 nm) (c=2, EtOH)

Specification Test Results

Related Information

Frequently Bought Together