Order Entry
Denmark
ContactUsLinkComponent
 
Theophyllin, anhydrous 99+%
 
undefined
Theophyllin, anhydrous 99+%
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    270 to 274 °C
  • Titration with NaOH:
    ≥99.0 %

 

 

Frequently Bought Together