Order Entry
Denmark
ContactUsLinkComponent
Colour disc test kit, chlorine dioxide
Colour disc test kit, chlorine dioxide
Catalog # HACH2842800
Supplier:  Hach
CAS Number:  
undefined
Colour disc test kit, chlorine dioxide
Catalog # HACH2842800
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    DPD
  • Pk:
    100 Tests
  • Description:
    Chlorine dioxide color disc test kit, model CLD-2
  • Seq:
    01
  • Range:
    0 - 2,0 mg/l

Specifications

About this item

The chlorine dioxide test kit uses the reliable and EPA-accepted DPD method to monitor ClO2 concentration, while eliminating chlorine interference through the addition of glycine.

  • Simple, cost-effective method for maintaining optimum chlorine dioxide levels in your plant
  • USEPA-accepted for waste water and potable water

With available range of 0 to 2 mg/l and resolution of 0,1 mg/l, operators can accurately monitor around the typical 0,8 mg/l regulated limit.

Delivery: Kit includes DPD powder pillows, glycine reagent, plastic viewing tube, colour comparator box, colour discs, instruction sheets, and carrying case.