Reference standards VOA
Supplier: Restek
About this item
38 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- REST30025
- REST30027
- REST30029
- REST30030
- REST30032
- REST30223
- REST30224
- REST30245
- REST30248
- REST30249
- REST30250
- REST30252
- REST30254
- REST30258
- REST30261
- REST30263
- REST30266
- REST30269
- REST30272
- REST30276
- REST30278
- REST30286
- REST30288
- REST30290
- REST30293
- REST30294
- REST30403
- REST30404
- REST30409
- REST30413
- REST30414
- REST30415
- REST30416
- REST30420
- REST30424
- REST30425
- REST30426
- REST30429
- REST30430
- Description:
- Benzene-d6, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,2-dichloroethane-d4, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Ethylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Fluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,4-difluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Chlorobenzene-d5, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Toluene-d8, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Acetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Allyl chloride, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Benzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Bromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Bromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 2-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Carbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Chlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Chloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Chloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 4-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,2-Dibromoethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,1-dichloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Diethyl ether, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Ethanol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Ethylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Isobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Isopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 2-nitropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Pentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Pyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Tetrachloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Tetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Toluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Trichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- m-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- o-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- p-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- 1,2,3-trichloropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Dibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- CAS No.:
- 3114-55-4
- 2037-26-5
- 67-64-1
- 107-05-1
- 71-43-2
- 108-86-1
- 75-25-2
- 78-93-3
- 75-15-0
- 108-90-7
- 75-00-3
- 67-66-3
- 106-43-4
- 106-93-4
- 75-34-3
- 75-35-4
- 60-29-7
- 64-17-5
- 100-41-4
- 78-83-1
- 98-82-8
- 79-46-9
- 76-01-7
- 110-86-1
- 127-18-4
- 109-99-9
- 108-88-3
- 87-61-6
- 79-01-6
- 108-38-3
- 95-47-6
- 106-42-3
- 96-18-4
- 74-95-3
- Formula:
- C₆H₆
- C₇H₈
- C₃H₆₀
- C₃H₅Cl
- C₆H₆
- C₆H₅Br
- CHBr₃
- C₄H₈O
- CS₂
- C₆H₅Cl
- C₂H₅Cl
- CHCl₃
- C₇H₇Cl
- C₂H₄Br₂
- C₂H₄Cl₂
- C₂H₂Cl₂
- C₄H₁₀O
- C₂H₆O
- C₈H₁₀
- C₄H₁₀O
- C₉H₁₂
- C₃H₇NO₂
- C₂HCl₅
- C₅H₅N
- C₂Cl₄
- C₄H₈O
- C₇H₈
- C₆H₃Cl₃
- C₂HCl₃
- C₈H₁₀
- C₈H₁₀
- C₈H₁₀
- C₃H₅Cl₃
- CH₂Br₂
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- REST30025Benzene-d6, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleBenzene-d6, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr407.00Specifications:
More
Specifications
Cat. No.REST30025DescriptionBenzene-d6, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.Formula - REST300271,2-dichloroethane-d4, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,2-dichloroethane-d4, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30027Description1,2-dichloroethane-d4, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.Formula - REST30029Ethylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleEthylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr407.00Specifications:
More
Specifications
Cat. No.REST30029DescriptionEthylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.Formula - REST30030Fluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleFluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30030DescriptionFluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.Formula - REST300321,4-difluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,4-difluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30032Description1,4-difluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.Formula - REST30223Chlorobenzene-d5, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleChlorobenzene-d5, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr422.00Specifications:
More
Specifications
Cat. No.REST30223DescriptionChlorobenzene-d5, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.3114-55-4FormulaC₆H₆ - REST30224Toluene-d8, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleToluene-d8, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr422.00Specifications:
More
Specifications
Cat. No.REST30224DescriptionToluene-d8, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.2037-26-5FormulaC₇H₈ - REST30245Acetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampuleAcetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr306.00Specifications:
More
Specifications
Cat. No.REST30245DescriptionAcetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.67-64-1FormulaC₃H₆₀ - REST30248Allyl chloride, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleAllyl chloride, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLSpecifications:
More
Specifications
Cat. No.REST30248DescriptionAllyl chloride, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.107-05-1FormulaC₃H₅Cl - REST30249Benzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleBenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30249DescriptionBenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.71-43-2FormulaC₆H₆ - REST30250Bromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleBromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr306.00Specifications:
More
Specifications
Cat. No.REST30250DescriptionBromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.108-86-1FormulaC₆H₅Br - REST30252Bromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleBromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr392.00Specifications:
More
Specifications
Cat. No.REST30252DescriptionBromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.75-25-2FormulaCHBr₃ - REST302542-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule2-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30254Description2-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.78-93-3FormulaC₄H₈O - REST30258Carbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCarbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr392.00Specifications:
More
Specifications
Cat. No.REST30258DescriptionCarbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.75-15-0FormulaCS₂ - REST30261Chlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleChlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr299.00Specifications:
More
Specifications
Cat. No.REST30261DescriptionChlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.108-90-7FormulaC₆H₅Cl - REST30263Chloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleChloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr422.00Specifications:
More
Specifications
Cat. No.REST30263DescriptionChloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.75-00-3FormulaC₂H₅Cl - REST30266Chloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleChloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30266DescriptionChloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.67-66-3FormulaCHCl₃ - REST302694-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule4-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Discontinued-Specifications:
More
Specifications
Cat. No.REST30269Description4-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.106-43-4FormulaC₇H₇Cl - REST302721,2-Dibromoethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,2-Dibromoethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30272Description1,2-Dibromoethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.106-93-4FormulaC₂H₄Br₂ - REST302761,1-dichloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,1-dichloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30276Description1,1-dichloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.75-34-3FormulaC₂H₄Cl₂ - REST302781,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr407.00Specifications:
More
Specifications
Cat. No.REST30278Description1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.75-35-4FormulaC₂H₂Cl₂ - REST30286Diethyl ether, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleDiethyl ether, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr280.00Specifications:
More
Specifications
Cat. No.REST30286DescriptionDiethyl ether, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.60-29-7FormulaC₄H₁₀O - REST30288Ethanol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleEthanol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30288DescriptionEthanol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.64-17-5FormulaC₂H₆O - REST30290Ethylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleEthylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30290DescriptionEthylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.100-41-4FormulaC₈H₁₀ - REST30293Isobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleIsobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30293DescriptionIsobutyl alcohol, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.78-83-1FormulaC₄H₁₀O - REST30294Isopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleIsopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30294DescriptionIsopropylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.98-82-8FormulaC₉H₁₂ - REST304032-nitropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule2-nitropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr299.00Specifications:
More
Specifications
Cat. No.REST30403Description2-nitropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.79-46-9FormulaC₃H₇NO₂ - REST30404Pentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampulePentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr280.00Specifications:
More
Specifications
Cat. No.REST30404DescriptionPentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.76-01-7FormulaC₂HCl₅ - REST30409Pyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampulePyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr392.00Specifications:
More
Specifications
Cat. No.REST30409DescriptionPyridine, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.110-86-1FormulaC₅H₅N - REST30413Tetrachloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleTetrachloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30413DescriptionTetrachloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.127-18-4FormulaC₂Cl₄ - REST30414Tetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleTetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr306.00Specifications:
More
Specifications
Cat. No.REST30414DescriptionTetrahydrofuran, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.109-99-9FormulaC₄H₈O - REST30415Toluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleToluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30415DescriptionToluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.108-88-3FormulaC₇H₈ - REST304161,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30416Description1,2,3-trichlorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.87-61-6FormulaC₆H₃Cl₃ - REST30420Trichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleTrichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr392.00Specifications:
More
Specifications
Cat. No.REST30420DescriptionTrichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.79-01-6FormulaC₂HCl₃ - REST30424m-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampulem-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30424Descriptionm-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.108-38-3FormulaC₈H₁₀ - REST30425o-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleo-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr396.00Specifications:
More
Specifications
Cat. No.REST30425Descriptiono-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.95-47-6FormulaC₈H₁₀ - REST30426p-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampulep-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 Itemskr396.00Specifications:
More
Specifications
Cat. No.REST30426Descriptionp-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.106-42-3FormulaC₈H₁₀ - REST304291,2,3-trichloropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1,2,3-trichloropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1 mLLog in for availability1 * 1 mLkr392.00Specifications:
More
Specifications
Cat. No.REST30429Description1,2,3-trichloropropane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.96-18-4FormulaC₃H₅Cl₃ - REST30430Dibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleDibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule1Log in for availability1 * 1 ItemsSpecifications:
More
Specifications
Cat. No.REST30430DescriptionDibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampuleCAS No.74-95-3FormulaCH₂Br₂
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



