Perchlorsyre 0.1 N in aqueous solution USP Test Solution (TS) 34 conforms to USP 34
Leverandør: VWR Chemicals
Certifikater
Om denne vare
CAS nummer:7601-90-3
:
Formel:HClO₄
MDL nummer:MFCD00011325
:100.46 g/mol
Opbevaringstemperatur:Stue temperatur
EINECS:0
FN:3264
ADR:8,III
:GHS05
:GHS03
Specifikationer
- Environmentally Preferable:
Specifikationer
Frequently Bought Together