HRP substrate, 1-Step™ Slow TMB-ELISA
Catalog # PIER34024
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mL
- Description:1-Step™ Slow TMB-ELISA
Specifications
About this item
The 1-Step™ Slow TMB-ELISA contains soluble TMB (3,3´,5,5´-tetramentylbenzidine) that is ideal for ELISA-based applications. TMB is a chromogenic substrate for horseradish peroxidase. Once oxidised by the enzyme, this substrate yields a blue product that absorbs at 370 nm and 652 nm. Adding sulfuric or phosphoric acid to stop the reaction changes the product to yellow that absorbs at 450 nm and provides greater sensitivity than the blue product. The 1-Step Slow TMB-ELISA is supplied ready to use with no mixing required.
- No additional reagents or filtering required
- Non carcinogenic
- No DMF or DMSO present