IgM purification kit , Pierce™
About this item
The Pierce™ IgM Purification Kit is an easy to use affinity matrix and buffer system to purify IgM from mouse ascites fluid. The method uses immobilised mannan binding protein (MBP), which binds to IgM in the kit-supplied buffers.
- Immobilised MBP allows specific purification of mouse IgM from ascites fluid
- Non denaturing, near-neutral buffer conditions ensure recovery of functional antibodies
- Stable ligand and resins can be regenerated and reused at least ten times
Delivery information: Kit includes MBP Agarose column, 5 m, 1 column; IgM Binding buffer, 800 ml; IgM Elution buffer, 500 ml; MBP Column Preparation buffer, 50 ml; and Column extender.
3 Options Available Below
Product Details & Documents
The Pierce™ IgM Purification Kit is an easy to use affinity matrix and buffer system to purify IgM from mouse ascites fluid. The method uses immobilised mannan binding protein (MBP), which binds to IgM in the kit-supplied buffers.
- Immobilised MBP allows specific purification of mouse IgM from ascites fluid
- Non denaturing, near-neutral buffer conditions ensure recovery of functional antibodies
- Stable ligand and resins can be regenerated and reused at least ten times
Delivery information: Kit includes MBP Agarose column, 5 m, 1 column; IgM Binding buffer, 800 ml; IgM Elution buffer, 500 ml; MBP Column Preparation buffer, 50 ml; and Column extender.
Product Family Options
Product Information
- PkAvailabilityPrice
- PIER44897Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samplesPierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples1 KITLog in for availability1 * 1 Each9.350,00 kr
- PIER21018MBP Column preparation buffer
- PIER22212MBP Agarose, mannan binding protein on 4% beaded agaroseMBP Agarose, mannan binding protein on 4% beaded agarose10 mLLog in for availability1 * 10 mL13.810,00 kr