PARP (cleaved) colorimetric in-cell ELISA Kit, multispecies, Pierce™
PIER62219
:
- Pk:1
- Host:
- Primary antibody reactivity:
- Description:PARP Colorimetric In-Cell ELISA Kit, 96 assays
- Environmentally Preferable:
- Tests per kit:96 assays
Cleaved PARP Colorimetric In-Cell ELISA kit is a near infrared microplate assay optimised to measure activated (cleaved) poly (ADP-ribose) polymerase (PARP) and tubulin in whole cells.
- Whole-cell detection requires no cell lysis, protein extraction or special sample preparation
- Provides better data, higher throughput and less cost than quantitative Western blotting
- For use with standard microplate readers
: The kit includes rabbit monoclonal anti-(cleaved) PARP antibody, mouse monoclonal anti-alpha-Tubulin antibody, blocking buffer 50 ml, 20X tris buffered saline 50 ml, Surfact-Amps 20 (10% detergent) 10 ml, Surfact-Amps X-100 (10% detergent) 10 ml, HRP conjugate 0,14 ml, TMB substrate 58 ml, TMB stop solution 55 ml, Janus Green whole-cell stain 50 ml, elution buffer 2×30 ml and thin plate seal assembly.