Order Entry
Denmark
ContactUsLinkComponent
 
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate ≥99%, powder
 
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate ≥99%, powder

 

  • Synonyms:
    AMP; 5'-Adenylic acid; 5′-AMP-Na2
  • Form:
    White Powder
  • Storage:
    Store at −0 °C.
  • Optical Rotation:
    [α]20/D −43± 2°, c = 1% in 0,5 M Na2HPO4 (dry matter) (Lit.)
  • pH:
    7 (Lit.)
  • Source:
    Yeast

 

Frequently Bought Together