1-Octadecanol NF
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
CAS Number:112-92-5
:
Formula:H₃C(CH₂)₁₆CH₂OH
MDL Number:MFCD00002823
:270.5 g/mol
Merck Index:12,08960
Boiling Pt:336 °C (1013 hPa)
Melting Pt:56…57 °C
Density:0.805…0.815 g/cm³ (60 °C)
Storage Temperature:Ambient
EINECS:204-017-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:90% Min.
Specification Test Results
Frequently Bought Together