Riboflavine (vitamin B2) ≥98.0% USP
- Assay (C₁₇H₂₀N₄O₆):98.0 - 102.0 %
- Specific Rotation [A]D:-115 to -135
- Loss On Drying:max. 1.5 %
Frequently Bought Together
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Jerricans, Wide Neck, HDPE, with Screw Caps
: VWR Collection
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.
: Thermo Fisher Scientific
Nicotinic acid (Niacin) 98.0-102.0% (dried basis), powder USP
: Spectrum Chemical