Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-(2-Ethoxyethoxy)ethyl acetate

Supplier:   Spectrum Chemical
2-(2-Ethoxyethoxy)ethyl acetate
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:–25 °C
  • Density:1.0114 g/cm³ (20 °C)
  • EINECS:203-940-1
Specifications for Product Family

Specification Test Results

  • Assay:
    99% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Steel pail
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L

  • 200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L - 26.340,00 kr
    26.340,00 kr
    1 * 200 L
       26.340,00 kr