Order Entry
Denmark
ContactUsLinkComponent
Dextran, M.W. ~ 150,000, powder
Dextran, M.W. ~ 150,000, powder
Catalog # SPCMD1013-1KG
Supplier:  Spectrum Chemical
CAS Number:  
Dextran, M.W. ~ 150,000, powder
Catalog # SPCMD1013-1KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
EINECS:232-677-5
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935