1-Hexadecanol NF
Supplier: Spectrum Chemical
About this item
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Boiling Pt:305…330 °C (1013 hPa)
- Melting Pt:46…52 °C
- Density:0.81 g/cm³ (20 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:253-149-0
2 Options Available Below
Product Details & Documents
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Boiling Pt:305…330 °C (1013 hPa)
- Melting Pt:46…52 °C
- Density:0.81 g/cm³ (20 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:253-149-0
Specifications for Product Family
Specification Test Results
- Assay:90% Min.



