Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Methoxyflurane 97%

5 g
Catalog # L17285.06
Flammable
Health
Irritant
Methoxyflurane 97%
Catalog # L17285.06
CAS Number:  76-38-0
Methoxyflurane 97%
Methoxyflurane 97%

5 g

CAS Number:  76-38-0
Price
1.020,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 76-38-0 Molecular Formula: CH3OCF2CHCl2 Formula Weight: 164.97 Flash Point: 37°(98°F) Melting Point: -36° Boiling Point: 105° Beilstein Reference: 1737766 UN No.: UN3271 MDL No.: MFCD00040144

Synonyms:  2,2-Dichloro-1,1-difluoroethyl methyl ether
  • CAS Number:76-38-0
  • Formula:C₃H₄Cl₂F₂O
  • MDL Number:MFCD00040144
  • Molecular Weight:164.961 g/mol
  • Merck Index:14,5994
  • Boiling Pt:105 °C
  • Melting Pt:–36 °C
  • Density:1.440
  • Flash Pt:37 °C (98 °F)
  • Storage Temperature:Ambient
  • EINECS:200-956-0
  • ADR:3,III
Specifications

Specifications

  • Pk:
    5 g

Specifications