Order Entry
Denmark
ContactUsLinkComponent
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone 98%
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone 98%
Catalog # L11919.MD
CAS Number:  
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone 98%
Catalog # L11919.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :114.10 g/mol
Melting Pt:34…43 °C
Storage Temperature:Ambient
CAS Number:78508-96-0
Formula:C₅H₆O₃
MDL Number:MFCD00075370