Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ticarcillin disodium : clavulanate potassium
Ticarcillin disodium : clavulanate potassium (15:1)
Ticarcillin disodium : clavulanate potassium (15:1)
Ticarcillin disodium : clavulanate potassium (15:1)
Catalog # J66659.09
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Expand image
Ticarcillin disodium : clavulanate potassium (15:1)
Catalog # J66659.09
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Specifications
Pk:
10 g
Seq:
3236
Specifications
About this item
Product Details
Documentation
:
:
GHS08
:
GHS07
Storage Temperature:
Refrigerator
Product Details
Documentation