Midecamycin 900 µg/mg
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35457-80-8
:
Formula:C₄₁H₆₇NO₁₅
MDL Number:MFCD00865000
:813.97 g/mol
Boiling Pt:874 °C
Melting Pt:155 …156 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Refrigerator
EINECS:252-578-0
Related Information
Frequently Bought Together