Order Entry
Denmark
ContactUsLinkComponent
Arabinofuranosyl-2-fluoroadenine 98%
Arabinofuranosyl-2-fluoroadenine 98%
Catalog # J65562.03
CAS Number:  
Arabinofuranosyl-2-fluoroadenine 98%
Catalog # J65562.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:21679-14-1
 :
Formula:C₁₀H₁₂FN₅O₄
MDL Number:MFCD00132942
 :285.24 g/mol
Merck Index:14,4126
EINECS:244-525-5