Order Entry
Denmark
ContactUsLinkComponent
Lysis buffer, 0,5% Tween 20, pH 8,0, for isolation of His-tag modified proteins
Lysis buffer, 0,5% Tween 20, pH 8,0, for isolation of His-tag modified proteins
Catalog # J61792.AP
CAS Number:  
Lysis buffer, 0,5% Tween 20, pH 8,0, for isolation of His-tag modified proteins
Catalog # J61792.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Lysis buffer, 0.5% Tween 20, pH 8.0, for isolation of His-tag modified proteins
  • MDL No.:
    MFCD00677267
  • Physical form:
    Liquid

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00677267
Applications: For isolation of His-tag proteins