3,3',5,5'-Tetramethylbenzidine (TMB) solution, high sensitivity ready-to-use
Catalog # J61325.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mL
Specifications
About this item
CAS Number:54827-17-7
:
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748
:240.35 g/mol
Storage Temperature:Refrigerator
EINECS:259-364-6
UN:2810
ADR:6.1,III
:GHS08
:GHS07