Order Entry
Denmark
ContactUsLinkComponent
Phosphate-buffered saline (PBS, 1X), sterile filtered
Phosphate-buffered saline (PBS, 1X), sterile filtered
Catalog # J61196.AP
CAS Number:  
Phosphate-buffered saline (PBS, 1X), sterile filtered
Catalog # J61196.AP
CAS Number:  

Specifications

  • Concentration:
    1X
  • Pk:
    500 mL
  • Description:
    Phosphate-buffered saline (PBS), sterile filtered
  • Physical form:
    Liquid

Specifications

About this item

Physical Form: Liquid